MilliporeSigma
  • Characterization of the Key Aroma Compounds in Traditional Hunan Smoke-Cured Pork Leg (Larou, THSL) by Aroma Extract Dilution Analysis (AEDA), Odor Activity Value (OAV), and Sensory Evaluation Experiments.
Characterization of the Key Aroma Compounds in Traditional Hunan Smoke-Cured Pork Leg (Larou, THSL) by Aroma Extract Dilution Analysis (AEDA), Odor Activity Value (OAV), and Sensory Evaluation Experiments.
Foods (Basel, Switzerland) (2020-04-08)
Dandan Pu, Yuyu Zhang, Huiying Zhang, Baoguo Sun, Fazheng Ren, Haitao Chen, Yizhuang Tang
ABSTRACT
The key aroma compounds in smoke-cured pork leg were characterized by gas chromatography-olfactometry coupled with aroma extract dilution analysis (GC-O/AEDA), odor activity value (OAV), recombination modeling, and omission tests. Ranking analysis showed that pork leg smoke-cured for 18 days had the best sensory qualities, with strong meaty, smoky, roasty, woody, and greasy attributes. Thirty-nine aroma-active regions with flavor dilution (FD) factors ranging from 9 to 6561 were detected. Overall, 3-ethylphenol had the highest FD factor of 6561, followed by 2,6-dimethoxyphenol, 3,4-dimethylphenol, 4-ethylguaiacol, 4-methylguaiacol, 3-methylphenol, and 2-acetyl-1-pyrroline, with FD ≥243. Among 39 aroma compounds, 27 compounds with OAVs ≥1 and were potent odorants. A similarity of 90.73% between the recombination model and traditional Hunan Smoke-cured Pork Leg (THSL) sample was obtained. Omission tests further confirmed that (E)-2-nonenal, 2-methoxy-4-vinylphenol, guaiacol, 3-ethylphenol, 2,6-dimethylphenol, 2-acetyl-1-pyrroline, and methional were key odorants in smoke-cured pork leg. Additionally, 2-acetyl-1-pyrroline (38.88 μg/kg), which contributes to a roasty aroma, was characterized here as a key odorant of smoke-cured pork leg for the first time.
Sigma-Aldrich
3,4,5-Trimethoxytoluene, 97%
Sigma-Aldrich
3,5-Dimethoxyphenol, 99%
Sigma-Aldrich
2-Methoxy-4-propylphenol, ≥99%, FG
Sigma-Aldrich
trans-2-Nonenal, 97%