MilliporeSigma
  • CARD3 Promotes Cerebral Ischemia-Reperfusion Injury Via Activation of TAK1.
CARD3 Promotes Cerebral Ischemia-Reperfusion Injury Via Activation of TAK1.
Journal of the American Heart Association (2020-05-01)
Xiaolin Wu, Lijin Lin, Juan-Juan Qin, Lifen Wang, Hao Wang, Yichun Zou, Xueyong Zhu, Ying Hong, Yan Zhang, Ye Liu, Can Xin, Shuangxiang Xu, Shengda Ye, Jianjian Zhang, Zhongwei Xiong, Lihua Zhu, Hongliang Li, Jincao Chen, Zhi-Gang She
ABSTRACT
Background Although multiple signaling cascades and molecules contributing to the pathophysiological process have been studied, the treatments for stroke against present targets have not acquired significant clinical progress. Although CARD3 (caspase activation and recruitment domain 3) protein is an important factor involved in regulating immunity, inflammation, lipid metabolism, and apoptosis, its role in cerebral stroke is currently unknown. Methods and Results Using a mouse model of ischemia-reperfusion (I-R) injury based on transient blockage of the middle cerebral artery, we have found that CARD3 expression is upregulated in a time-dependent manner during I-R injury. Further animal study revealed that, relative to control mice, CARD3-knockout mice exhibited decreased inflammatory response and neuronal apoptosis, with reduced infarct volume and lower neuropathological scores. In contrast, neuron-specific CARD3-overexpressing transgenic (CARD3-TG) mice exhibited increased I-R induced injury compared with controls. Mechanistically, we also found that the activation of TAK1 (transforming growth factor-β-activated kinase 1) was enhanced in CARD3-TG mice. Furthermore, the increased inflammation and apoptosis seen in injured CARD3-TG brains were reversed by intravenous administration of the TAK1 inhibitor 5Z-7-oxozeaenol. Conclusions These results indicate that CARD3 promotes I-R injury via activation of TAK1, which not only reveals a novel regulatory axis of I-R induced brain injury but also provides a new potential therapeutic approach for I-R injury.
Sigma-Aldrich
5Z-7-Oxozeaenol, ≥98% (HPLC)
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
ApopTag Plus In Situ Apoptosis Fluorescein Detection Kit, The ApopTag Plus Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a fluorescein reporter molecule.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
Anti-NeuN Antibody, from chicken, purified by affinity chromatography