MilliporeSigma
  • Structural Elucidation of Malonylcommunol and 6β-Hydroxy-trans-communic Acid, Two Undescribed Diterpenes from Salvia cinnabarina. First Examples of Labdane Diterpenoids from a Mexican Salvia Species.
Structural Elucidation of Malonylcommunol and 6β-Hydroxy-trans-communic Acid, Two Undescribed Diterpenes from Salvia cinnabarina. First Examples of Labdane Diterpenoids from a Mexican Salvia Species.
Molecules (Basel, Switzerland) (2020-04-25)
Celia Bustos-Brito, Antonio Nieto-Camacho, Simón Hernandez-Ortega, José Rivera-Chávez, Leovigildo Quijano, Baldomero Esquivel
ABSTRACT
The aerial parts of Salvia cinnabarina afforded two undescribed labdane diterpenoids 1 and 2 (malonylcommunol and 6β-hydroxy-trans-communic acid) along with two known labdane diterpenoids, trans-communic acid (3) and trans-communol (4). Additionally, seven known metabolites were also isolated; two isopimarane diterpenoids 5 and 6, two sesquiterpenoids identified as β-eudesmol (7) and cryptomeridiol (8), and three aromatic compounds identified as phthalic acid (9), a mixture of tyrosol fatty acid esters (10) and the flavone salvigenine (11). While compounds compounds 1-3 showed significant inhibition of yeast α-glucosidase, compounds 2, 3 and 7 had no anti-inflammatory activity in the edema model induced by TPA. This paper is not only the first report on a wild population of Salvia cinnabarina, but also of the presence of labdane-type diterpenoids in a Mexican Salvia sp.
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, Type I, lyophilized powder, ≥10 units/mg protein (using p-nitrophenyl α-D-glucoside as substrate.)
Pricing and availability is not currently available.
Sigma-Aldrich
4-Nitrophenyl α-D-glucopyranoside, ≥99%
Pricing and availability is not currently available.