MilliporeSigma
The SUMOylated METTL8 Induces R-loop and Tumorigenesis via m3C.
iScience (2020-03-22)
Li-Hong Zhang, Xue-Yun Zhang, Tao Hu, Xin-Yun Chen, Jing-Jia Li, Manfred Raida, Ning Sun, Yan Luo, Xiang Gao
ABSTRACT
R-loops, three-stranded DNA-DNA:RNA hybrid structures, are best known for their deleterious effects on genome stability. The regulatory factors of this fundamental genetic structure remain unclear. Here, we reveal an epigenetic factor that controls R-loop stability. METTL8, a member of the methyltransferase-like protein family that methylates 3-methylcytidine (m3C), is a key factor in the R-loop regulating methyltransferase complex. Biochemical studies show that METTL8 forms a large SUMOylated nuclear RNA-binding protein complex (∼0.8 mega daltons) that contains well-reported R-loop related factors. Genetic ablation of METTL8 results in an overall reduction of R-loops in cells. Interaction assays indicated METTL8 binds to RNAs and is responsible for R-loop stability on selected gene regions. Our results demonstrate that the SUMOylated METTL8 promotes tumorigenesis by affecting genetic organization primarily in, or in close proximity to, the nucleolus and impacts the formation of regulatory R-loops through its methyltransferase activity on m3C.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Minimum Essential Medium Eagle, Joklik Modification, with L-glutamine, without calcium chloride and sodium bicarbonate, suitable for cell culture
Sigma-Aldrich
Doxycycline hydrochloride
Corning® ProCulture® glass spinner flask complete, baffled, angled sidearms, 125 mL glass spinner flask, w/ 70 mm flat center cap, 2 32 mm vertical sidearms, reusable, non-sterile, 1/cs