MilliporeSigma
  • Selecting for Altered Substrate Specificity Reveals the Evolutionary Flexibility of ATP-Binding Cassette Transporters.
Selecting for Altered Substrate Specificity Reveals the Evolutionary Flexibility of ATP-Binding Cassette Transporters.
Current biology : CB (2020-03-30)
Sriram Srikant, Rachelle Gaudet, Andrew W Murray
ABSTRACT
ATP-binding cassette (ABC) transporters are the largest family of ATP-hydrolyzing transporters, which import or export substrates across membranes, and have members in every sequenced genome. Structural studies and biochemistry highlight the contrast between the global structural similarity of homologous transporters and the enormous diversity of their substrates. How do ABC transporters evolve to carry such diverse molecules and what variations in their amino acid sequence alter their substrate selectivity? We mutagenized the transmembrane domains of a conserved fungal ABC transporter that exports a mating pheromone and selected for mutants that export a non-cognate pheromone. Mutations that alter export selectivity cover a region that is larger than expected for a localized substrate-binding site. Individual selected clones have multiple mutations, which have broadly additive contributions to specific transport activity. Our results suggest that multiple positions influence substrate selectivity, leading to alternative evolutionary paths toward selectivity for particular substrates and explaining the number and diversity of ABC transporters.
Product Description
Millipore
PVDF Membrane Filter, 0.22 μm Pore Size, Durapore®, filter diam. 25 mm, hydrophilic
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)