MilliporeSigma
  • Wounding Induces Facultative Opn5-Dependent Circadian Photoreception in the Murine Cornea.
Wounding Induces Facultative Opn5-Dependent Circadian Photoreception in the Murine Cornea.
Investigative ophthalmology & visual science (2020-06-17)
Nicolás M Díaz, Richard A Lang, Russell N Van Gelder, Ethan D Buhr
ABSTRACT
Autonomous molecular circadian clocks are present in the majority of mammalian tissues. These clocks are synchronized to phases appropriate for their physiologic role by internal systemic cues, external environmental cues, or both. The circadian clocks of the in vivo mouse cornea synchronize to the phase of the brain's master clock primarily through systemic cues, but ex vivo corneal clocks entrain to environmental light cycles. We evaluated the underlying mechanisms of this difference. Molecular circadian clocks of mouse corneas were evaluated in vivo and ex vivo for response to environmental light. The presence of opsins and effect of genetic deletion of opsins were evaluated for influence on circadian photoresponses. Opn5-expressing cells were identified using Opn5Cre;Ai14 mice and RT-PCR, and they were characterized using immunocytochemistry. Molecular circadian clocks of the cornea remain in phase with behavioral circadian locomotor rhythms in vivo but are photoentrainable in tissue culture. After full-thickness incision or epithelial debridement, expression of the opsin photopigment Opn5 is induced in the cornea in a subset of preexisting epithelial cells adjacent to the wound site. This induction coincides with conferral of direct, short-wavelength light sensitivity to the circadian clocks throughout the cornea. Corneal circadian rhythms become photosensitive after wounding. Opn5 gene function (but not Opn3 or Opn4 function) is necessary for induced photosensitivity. These results demonstrate that opsin-dependent direct light sensitivity can be facultatively induced in the murine cornea.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
(±)-2-Amino-4-phosphonobutyric acid, solid