MilliporeSigma
  • B7-H3-Targeted CAR-T Cells Exhibit Potent Antitumor Effects on Hematologic and Solid Tumors.
B7-H3-Targeted CAR-T Cells Exhibit Potent Antitumor Effects on Hematologic and Solid Tumors.
Molecular therapy oncolytics (2020-04-30)
Zongliang Zhang, Caiying Jiang, Zhiyong Liu, Meijia Yang, Xin Tang, Yuelong Wang, Meijun Zheng, Jianhan Huang, Kunhong Zhong, Shasha Zhao, Mei Tang, Tingyue Zhou, Hui Yang, Gang Guo, Liangxue Zhou, Jianguo Xu, Aiping Tong
ABSTRACT
Recently, B7-H3 was frequently reported to be overexpressed in various cancer types and has been suggested to be a promising target for cancer immunotherapy. In the present study, we analyzed the mRNA expression of B7-H3 in The Cancer Genome Atlas (TCGA) database and validated its expression across multiple cancer types. We then generated a novel B7-H3-targeted chimeric antigen receptor (CAR) and tested its antitumor activity both in vitro and in vivo. The B7-H3 expression heterogeneity and variation were frequent. Moderate or even high expression levels of B7-H3 were also observed in some tumor-adjacent tissues, but the staining intensity was weaker than that in tumor tissues. B7-H3 expression was absent or very low in normal tissues and organs. Flow cytometry indicated that the mean expression level of B7-H3 in eight bone marrow specimens from patients with acute myeloid leukemia (AML) was 57.2% (range 38.8-80.4). Furthermore, we showed that the B7-H3-targeted CAR-T cells exhibited significant antitumor activity against AML and melanoma in vitro and in xenograft mouse models. In conclusion, although B7-H3 represents a promising pan-cancer target, and B7-H3-redirected CAR-T cells can effectively control tumor growth, the expression heterogeneity and variation have to be carefully considered in translating B7-H3-targeted CAR-T cell therapy into clinical practice.
Sigma-Aldrich
CD276 human, recombinant, expressed in E. coli, 0.5 mg protein/mL