MilliporeSigma
  • Increased glycosidase activities improved the production of wine varietal odorants in mixed fermentation of P. fermentans and high antagonistic S. cerevisiae.
Increased glycosidase activities improved the production of wine varietal odorants in mixed fermentation of P. fermentans and high antagonistic S. cerevisiae.
Food chemistry (2020-07-04)
Na Li, Qian-Qian Wang, Yin-Hu Xu, Ai-Hua Li, Yong-Sheng Tao
ABSTRACT
A selected Pichia fermentans strain was simultaneously and sequentially inoculated in synthetic and real juice with S. cerevisiae strains of different antagonistic activities in a ratio 1:1 to observe the correlation between varietal odorants and glycosidase activities. Fermentations using pure S. cerevisiae strains were used for comparison. Yeast biomass and glycosidase activities were monitored, varietal odorants were detected using HS-SPME-GC/MS during fermentation. The final wine aroma attributes were analyzed by trained panelists. Results showed that co-inoculation with high antagonistic S. cerevisiae resulted in higher glycosidase activities than others. Pearson correlation analysis indicated that yeast biomass was positively related to glycosidase activities during fermentation. The increase in glycosidase activities was the main reason for the higher production of terpenes and C13-norisoprenoids, and for the lower C6 compound content, which lead to superior fruity and floral aromas in the final wine samples of the high antagonistic S. cerevisiae group.
Product Description
Supelco
SPME Fiber Holder, for use with Manual Sampling, pk of 1 ea