MilliporeSigma
3D printable tough silicone double networks.
Nature communications (2020-08-12)
Thomas J Wallin, Leif-Erik Simonsen, Wenyang Pan, Kaiyang Wang, Emmanuel Giannelis, Robert F Shepherd, Yiğit Mengüç
ABSTRACT
Additive manufacturing permits innovative soft device architectures with micron resolution. The processing requirements, however, restrict the available materials, and joining chemically dissimilar components remains a challenge. Here we report silicone double networks (SilDNs) that participate in orthogonal crosslinking mechanisms-photocurable thiol-ene reactions and condensation reactions-to exercise independent control over both the shape forming process (3D printing) and final mechanical properties. SilDNs simultaneously possess low elastic modulus (E100% < 700kPa) as well as large ultimate strains (dL/L0 up to ~ 400 %), toughnesses (U ~ 1.4 MJ·m-3), and strengths (σ ~ 1 MPa). Importantly, the latent condensation reaction permits cohesive bonding of printed objects to dissimilar substrates with modulus gradients that span more than seven orders of magnitude. We demonstrate soft devices relevant to a broad range of disciplines: models that simulate the geometries and mechanical properties of soft tissue systems and multimaterial assemblies for next generation wearable devices and robotics.
Sigma-Aldrich
3-(Trimethoxysilyl)propyl acrylate, 92%, contains 100 ppm BHT as inhibitor
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)