MilliporeSigma
  • Incorporation of Pseudo-complementary Bases 2,6-Diaminopurine and 2-Thiouracil into Serinol Nucleic Acid (SNA) to Promote SNA/RNA Hybridization.
Incorporation of Pseudo-complementary Bases 2,6-Diaminopurine and 2-Thiouracil into Serinol Nucleic Acid (SNA) to Promote SNA/RNA Hybridization.
Chemistry, an Asian journal (2020-02-06)
Yukiko Kamiya, Fuminori Sato, Keiji Murayama, Atsuji Kodama, Susumu Uchiyama, Hiroyuki Asanuma
ABSTRACT
Serinol nucleic acid (SNA) is a promising candidate for nucleic acid-based molecular probes and drugs due to its high affinity for RNA. Our previous work revealed that incorporation of 2,6-diaminpurine (D), which can form three hydrogen bonds with uracil, into SNA increases the melting temperature of SNA-RNA duplexes. However, D incorporation into short self-complementary regions of SNA promoted self-dimerization and hindered hybridization with RNA. Here we synthesized a SNA monomer of 2-thiouracil (sU), which was expected to inhibit base pairing with D by steric hindrance between sulfur and the amino group. To prepare the SNA containing D and sU in high yield, we customized the protecting groups on D and sU monomers that can be readily deprotected under acidic conditions. Incorporation of D and sU into SNA facilitated stable duplex formation with target RNA by suppressing the self-hybridization of SNA and increasing the stability of the heteroduplex of SNA and its complementary RNA. Our results have important implications for the development of SNA-based probes and nucleic acid drugs.
Product Description
Supelco
Acetic acid – triethylamine solution 1:1, suitable for HPLC, 2M:2M aqueous solution, LiChropur