MilliporeSigma
  • CircHIPK3 promotes proliferation and metastasis and inhibits apoptosis of renal cancer cells by inhibiting MiR-485-3p.
CircHIPK3 promotes proliferation and metastasis and inhibits apoptosis of renal cancer cells by inhibiting MiR-485-3p.
Cancer cell international (2020-06-20)
Jinjin Lai, Jun Xin, Changde Fu, Wei Zhang
ABSTRACT
The intervention of circHIPK3 in renal carcinoma (RC) has not been reported, and thus, the current study investigated the intervention and mechanism of circHIPK3 in RC. The expression of circHIPK3 in RC tissues and cells was detected by quantitative real-time polymerase chain reaction (qRT-PCR). Ribonuclease R (RNase R) resistance and distribution of circHIPK3 and HIPK3 were analyzed by RNase R digestion experiments and cytoplasm/nucleus separation experiments. CircHIPK3 was knocked down in ACHN and 769-P cells. Cell counting kit-8 (CCK-8), colony formation assay, scratch assay, and Transwell assay were performed to detect cell proliferation and metastasis. CircInteractome, qRT-PCR and dual-luciferase reporter assay were used to predict the target miRNAs of circHIPK3. Furthermore, a series of rescue experiments were performed to analyze the regulatory relationship between circHIPK3 and miR-485-3p. Epithelial-mesenchymal transition (EMT) and the expressions of apoptosis-associated markers were detected by Western blot and qRT-PCR. The regulatory relationship between circHIPK3 and miR-485-3p in vivo was explored by xenograft experiments, Western blot, qRT-PCR and immunohistochemistry (Ki-67). CircHIPK3 was mainly overexpressed in the cytoplasm of RC tissues and cells. Knocking down circHIPK3 inhibited the proliferation, migration, and invasion of RC cells. The expression of circHIPK3 was negatively related to that of its target gene miR-485-3p. Results of the rescue experiments showed that circHIPK3 overexpression could partially reverse the anti-carcinoma effect of miR-485-3p mimic. The specific mechanism of circHIPK3 was related to the effect of miR-485-3p on partially reversing the up-regulated expressions of Clever caspase-3, Bax, E-Cadherin and down-regulated expressions of Bcl-2, N-Cadherin and Vimentin. The results of in vivo experiments demonstrated that circHIPK3 promoted tumor growth and the expression of Ki-67 by down-regulating miR-485-3p. CircHIPK3 promotes the proliferation and metastasis and inhibits the apoptosis of RC cells through competitively binding to miR-485-3p.
Sigma-Aldrich
Giemsa Stain, Modified Solution, according to Giemsa
Millipore
Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein