MilliporeSigma
  • Oleic acid promotes cell invasion through an integrin-linked kinase signaling pathway in renal cell carcinoma.
Oleic acid promotes cell invasion through an integrin-linked kinase signaling pathway in renal cell carcinoma.
Nutrition and cancer (2019-10-02)
Zhen-Hua Liu, Cai-Peng Qing, Zheng-Zuo Sheng, Gui-Zhong Li, Li-Bo Man, Tao Xu
ABSTRACT
The change of fatty acid composition has been regarded as an indicator of altered lipid metabolism during human tumourigenesis, but the details are still unclear. We have previously demonstrated a monounsaturated fatty acid (MUFA) named oleic acid (OA) was involved in renal cell carcinoma (RCC) cell growth, as an extracellular signaling molecule to regulate 786-O cell proliferation via the integrin-linked kinase (ILK) pathway. In this study, we further observe the effects of OA on cell invasion of RCC and the potential mechanism by which OA worked was determined. The transwell invasion assay showed OA increased cell invasion of RCC in a dose-dependent manner. Western blotting results indicated ILK, COX-2, and MMP-9 proteins were involved for their high expressions and these effects were reversed when down-regulating the expression of ILK by special siRNA. The MMPs inhibitor GM6001 could weaken the abilities of OA on RCC cells invasion. These results suggested MUFA indeed affected cell invasion of RCC, which was depended by the regulation of ILK pathway.
Sigma-Aldrich
MISSION® esiRNA, targeting human ILK