MilliporeSigma
  • Characterization of Streptomyces piniterrae sp. nov. and Identification of the Putative Gene Cluster Encoding the Biosynthesis of Heliquinomycins.
Characterization of Streptomyces piniterrae sp. nov. and Identification of the Putative Gene Cluster Encoding the Biosynthesis of Heliquinomycins.
Microorganisms (2020-04-05)
Xiaoxin Zhuang, Zhiyan Wang, Chenghui Peng, Can Su, Congting Gao, Yongjiang Wang, Shengxiong Huang, Chongxi Liu
ABSTRACT
A novel actinomycete producing heliquinomycin and 9'-methoxy-heliquinomycin, designated strain jys28T, was isolated from rhizosphere soil of Pinus yunnanensis and characterized using a polyphasic approach. The strain had morphological characteristics and chemotaxonomic properties identical to those of members of the genus Streptomyces. It formed spiral chains of spores with spiny surfaces. The menaquinones detected were MK-9(H6), MK-9(H8) and MK-9(H4). The major fatty acids were iso-C16:0, C15:0, C16:1ω7с and anteiso-C15:0. The phospholipids were diphosphatidylglycerol, phosphatidylmethylethanolamine, phosphatidylethanolamine and phosphatidylinositol mannoside. The DNA G + C content of the draft genome sequence, consisting of 8.5 Mbp, was 70.6%. Analysis of the 16S rRNA gene sequence showed that strain jys28T belongs to the genus Streptomyces with the highest sequence similarities to Streptomyces chattanoogensis NBRC 13058T (99.2%) and Streptomyces lydicus DSM 40002T (99.2%) and phylogenetically clustered with them. Multilocus sequence analysis based on five other house-keeping genes (atpD, gyrB, rpoB, recA and trpB) and the low level of DNA-DNA relatedness and phenotypic differences allowed the novel isolate to be differentiated from its most closely related strains. Therefore, the strain is concluded to represent a novel species of the genus Streptomyces, for which the name Streptomycespiniterrae sp. nov. is proposed. Furthermore, the putative biosynthetic gene cluster of heliquinomycins was identified and the biosynthetic pathway was discussed. The type strain is jys28T (=CCTCC AA 2018051T =DSM 109823T).
Sigma-Aldrich
Kaurenoic acid, ≥95% (LC/MS-ELSD)