MilliporeSigma
  • Genetically distinct Group B Streptococcus strains induce varying macrophage cytokine responses.
Genetically distinct Group B Streptococcus strains induce varying macrophage cytokine responses.
PloS one (2019-09-20)
Rebecca A Flaherty, Elena C Borges, Jessica A Sutton, David M Aronoff, Jennifer A Gaddy, Margaret G Petroff, Shannon D Manning
ABSTRACT
Group B Streptococcus (GBS) is an opportunistic pathogen that causes preterm birth and neonatal disease. Although GBS is known to exhibit vast diversity in virulence across strains, the mechanisms of GBS-associated pathogenesis are incompletely understood. We hypothesized that GBS strains of different genotypes would vary in their ability to elicit host inflammatory responses, and that strains associated with neonatal disease would induce different cytokine profiles than those associated with colonization. Using a multiplexed, antibody-based protein detection array, we found that production of a discrete number of inflammatory mediators by THP-1 macrophage-like cells was universally induced in response to challenge with each of five genetically distinct GBS isolates, while other responses appeared to be strain-specific. Key array responses were validated by ELISA using the initial five strains as well as ten additional strains with distinct genotypic and phenotypic characteristics. Interestingly, IL-6 was significantly elevated following infection with neonatal infection-associated sequence type (ST)-17 strains and among strains possessing capsule (cps) type III. Significant differences in production of IL1-β, IL-10 and MCP-2 were also identified across STs and cps types. These data support our hypothesis and suggest that unique host innate immune responses reflect strain-specific differences in virulence across GBS isolates. Such data might inform the development of improved diagnostic or prognostic strategies against invasive GBS infections.
Sigma-Aldrich
Human MCP-2 / CCL8 ELISA Kit, for serum, plasma, cell culture supernatant and urine