MilliporeSigma
Context-Dependent Decision Making in a Premotor Circuit.
Neuron (2020-02-28)
Zheng Wu, Ashok Litwin-Kumar, Philip Shamash, Alexei Taylor, Richard Axel, Michael N Shadlen
ABSTRACT
Cognitive capacities afford contingent associations between sensory information and behavioral responses. We studied this problem using an olfactory delayed match to sample task whereby a sample odor specifies the association between a subsequent test odor and rewarding action. Multi-neuron recordings revealed representations of the sample and test odors in olfactory sensory and association cortex, which were sufficient to identify the test odor as match or non-match. Yet, inactivation of a downstream premotor area (ALM), but not orbitofrontal cortex, confined to the epoch preceding the test odor led to gross impairment. Olfactory decisions that were not context-dependent were unimpaired. Therefore, ALM does not receive the outcome of a match/non-match decision from upstream areas. It receives contextual information-the identity of the sample-to establish the mapping between test odor and action. A novel population of pyramidal neurons in ALM layer 2 may mediate this process.
Sigma-Aldrich
(+)-α-Pinene, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl butyrate, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
(R)-(+)-Limonene, 97%
Pricing and availability is not currently available.