MilliporeSigma
  • TFEB regulates murine liver cell fate during development and regeneration.
TFEB regulates murine liver cell fate during development and regeneration.
Nature communications (2020-05-20)
Nunzia Pastore, Tuong Huynh, Niculin J Herz, Alessia Calcagni', Tiemo J Klisch, Lorenzo Brunetti, Kangho Ho Kim, Marco De Giorgi, Ayrea Hurley, Annamaria Carissimo, Margherita Mutarelli, Niya Aleksieva, Luca D'Orsi, William R Lagor, David D Moore, Carmine Settembre, Milton J Finegold, Stuart J Forbes, Andrea Ballabio
ABSTRACT
It is well established that pluripotent stem cells in fetal and postnatal liver (LPCs) can differentiate into both hepatocytes and cholangiocytes. However, the signaling pathways implicated in the differentiation of LPCs are still incompletely understood. Transcription Factor EB (TFEB), a master regulator of lysosomal biogenesis and autophagy, is known to be involved in osteoblast and myeloid differentiation, but its role in lineage commitment in the liver has not been investigated. Here we show that during development and upon regeneration TFEB drives the differentiation status of murine LPCs into the progenitor/cholangiocyte lineage while inhibiting hepatocyte differentiation. Genetic interaction studies show that Sox9, a marker of precursor and biliary cells, is a direct transcriptional target of TFEB and a primary mediator of its effects on liver cell fate. In summary, our findings identify an unexplored pathway that controls liver cell lineage commitment and whose dysregulation may play a role in biliary cancer.
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %