MilliporeSigma
Extreme Drug Tolerance of Mycobacterium abscessus "Persisters".
Frontiers in microbiology (2020-03-21)
Yee-Kuen Yam, Nadine Alvarez, Mei-Lin Go, Thomas Dick
ABSTRACT
Persistence of infection despite extensive chemotherapy with antibiotics displaying low MICs is a hallmark of lung disease caused by Mycobacterium abscessus (Mab). Thus, the classical MIC assay is a poor predictor of clinical outcome. Discovery of more efficacious antibiotics requires more predictive in vitro potency assays. As a mycobacterium, Mab is an obligate aerobe and a chemo-organo-heterotroph - it requires oxygen and organic carbon sources for growth. However, bacteria growing in patients can encounter micro-environmental conditions that are different from aerated nutrient-rich broth used to grow planktonic cultures for MIC assays. These in vivo conditions may include oxygen and nutrient limitation which should arrest growth. Furthermore, Mab was shown to grow as biofilms in vivo. Here, we show Mab Bamboo, a clinical isolate we use for Mab drug discovery, can survive oxygen deprivation and nutrient starvation for extended periods of time in non-replicating states and developed an in vitro model where the bacterium grows as biofilm. Using these culture models, we show that non-replicating or biofilm-growing bacteria display tolerance to clinically used anti-Mab antibiotics, consistent with the observed persistence of infection in patients. To demonstrate the utility of the developed "persister" assays for drug discovery, we determined the effect of novel agents targeting membrane functions against these physiological forms of the bacterium and find that these compounds show "anti-persister" activity. In conclusion, we developed in vitro "persister" assays to fill an assay gap in Mab drug discovery compound progression and to enable identification of novel lead compounds showing "anti-persister" activity.
Sigma-Aldrich
ECO TWEEN® 80, viscous liquid
Supelco
Imipenem, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
TWEEN® 80, viscous liquid