MilliporeSigma
  • Activation of interferon regulatory factor 7 in plasmacytoid dendritic cells promotes experimental autoimmune pancreatitis.
Activation of interferon regulatory factor 7 in plasmacytoid dendritic cells promotes experimental autoimmune pancreatitis.
Journal of gastroenterology (2020-01-22)
Kosuke Minaga, Tomohiro Watanabe, Yasuyuki Arai, Masahiro Shiokawa, Akane Hara, Tomoe Yoshikawa, Ken Kamata, Kouhei Yamashita, Masatoshi Kudo
ABSTRACT
Excessive type I IFN (IFN-I) production by plasmacytoid dendritic cells (pDCs) promotes autoimmunity. Recently, we reported that a prominent feature of both experimental autoimmune pancreatitis (AIP) and human type 1 AIP is pDC activation followed by enhanced production of IFN-I and IL-33. However, the roles played by interferon regulatory factor 7 (IRF7), a critical transcription factor for IFN-I production in pDCs, in these disorders have not been clarified. Whole and nuclear extracts were isolated from pancreatic mononuclear cells (PMNCs) from MRL/MpJ mice exhibiting AIP. Expression of phospho-IRF7 and nuclear translocation of IRF7 was examined in these extracts by immunoblotting. Pancreatic expression of IRF7 was assessed by immunofluorescence analysis in experimental AIP. Nuclear translocation of IRF7 upon exposure to neutrophil extracellular traps (NETs) was assessed in peripheral blood pDCs from type 1 AIP patients. Pancreatic IRF7 expression was examined in surgically operated specimens from type 1 AIP patients. IRF7 activation was induced in pancreatic pDCs in experimental AIP. siRNA-mediated knockdown of IRF7 expression prevented AIP development, which was accompanied by a marked reduction in both pancreatic accumulation of pDCs and production of IFN-α and IL-33. Notably, in peripheral blood pDCs isolated from patients with type 1 AIP, nuclear translocation of IRF7 was enhanced as compared with the translocation in pDCs from healthy controls. Furthermore, IRF7-expressing pDCs were detected in the pancreas of patients with type 1 AIP. These findings suggest that the IRF7-IFN-I-IL-33 axis activated in pDCs drives pathogenic innate immune responses associated with type 1 AIP.
Sigma-Aldrich
MISSION® esiRNA, targeting human IRF7