MilliporeSigma
  • Carboxy-Terminal Phosphoregulation of the Long Splice Isoform of Free-Fatty Acid Receptor-4 Mediates β-Arrestin Recruitment and Signaling to ERK1/2.
Carboxy-Terminal Phosphoregulation of the Long Splice Isoform of Free-Fatty Acid Receptor-4 Mediates β-Arrestin Recruitment and Signaling to ERK1/2.
Molecular pharmacology (2020-03-07)
Ilya S Senatorov, Ameneh Cheshmehkani, Rebecca N Burns, Kirti Singh, Nader H Moniri
ABSTRACT
Free-fatty acid receptor-4 (FFA4), previously termed GPR120, is a G protein-coupled receptor (GPCR) for medium and long-chained fatty acids, agonism of which can regulate a myriad of metabolic, sensory, inflammatory, and proliferatory signals. Two alternative splice isoforms of FFA4 exist that differ by the presence of an additional 16 amino acids in the longer (FFA4-L) transcript, which has been suggested to be an intrinsically β-arrestin-biased GPCR. Although the shorter isoform (FFA4-S) has been studied more extensively, very little is known about mechanisms of regulation or signaling of the longer isoform. Because β-arrestin recruitment is dependent on receptor phosphorylation, in the current study, we used the endogenous agonist docosahexaenoic acid (DHA) to examine the mechanisms of FFA4-L phosphorylation, as well as DHA-dependent β-arrestin recruitment and DHA-dependent extracellular-signal regulated kinase-1/2 (ERK1/2) signaling in human embryonic kidney 293 cells. Our results reveal differences in basal phosphorylation of the two FFA4 isoforms, and we show that DHA-mediated phosphorylation of FFA4-L is primarily regulated by G protein-coupled receptor kinase 6, whereas protein kinase-C can also contribute to agonist-induced and heterologous phosphorylation. Moreover, our data demonstrate that FFA4-L phosphorylation occurs on the distal C terminus and is directly responsible for recruitment and interactions with β-arrestin-2. Finally, using CRISPR/Cas9 genome-edited cells, our data reveal that unlike FFA4-S, the longer isoform is unable to facilitate phosphorylation of ERK1/2 in cells that are devoid of β-arrestin-1/2. Together, these results are the first to demonstrate phosphoregulation of FFA4-L as well as the effects of loss of phosphorylation sites on β-arrestin recruitment and ERK1/2 activation. SIGNIFICANCE STATEMENT: Free-fatty acid receptor-4 (FFA4) is a cell-surface G protein-coupled receptor for medium and long-chained fatty acids that can be expressed as distinct short (FFA4-S) or long (FFA4-L) isoforms. Although much is known about FFA4-S, the longer isoform remains virtually unstudied. Here, we reveal the mechanisms of docosahexaenoic acid-induced phosphorylation of FFA4-L and subsequent β-arrestin-2 recruitment and extracellular-signal regulated kinase-1/2 activity.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Gq/11α Antibody, CT, Upstate®, from rabbit
Sigma-Aldrich
Bisindolylmaleimide II, ≥97% (Mixture of 2 isomers)