MilliporeSigma
  • Mannosylated Poly(ethylene imine) Copolymers Enhance saRNA Uptake and Expression in Human Skin Explants.
Mannosylated Poly(ethylene imine) Copolymers Enhance saRNA Uptake and Expression in Human Skin Explants.
Biomacromolecules (2020-04-07)
Anna K Blakney, Yamin Abdouni, Gokhan Yilmaz, Renjie Liu, Paul F McKay, Clément R Bouton, Robin J Shattock, C Remzi Becer
ABSTRACT
Messenger RNA (mRNA) is a promising platform for both vaccines and therapeutics, and self-amplifying RNA (saRNA) is particularly advantageous, as it enables higher protein expression and dose minimization. Here, we present a delivery platform for targeted delivery of saRNA using mannosylated poly(ethylene imine) (PEI) enabled by the host-guest interaction between cyclodextrin and adamantane. We show that the host-guest complexation does not interfere with the electrostatic interaction with saRNA and observed that increasing the degree of mannosylation inhibited transfection efficiency in vitro, but enhanced the number of cells expressing GFP by 8-fold in human skin explants. Besides, increasing the ratio of glycopolymer to saRNA also enhanced the percentage of transfected cells ex vivo. We identified that these mannosylated PEIs specifically increased protein expression in the epithelial cells resident in human skin in a mannose-dependent manner. This platform is promising for further study of glycosylation of PEI and targeted saRNA delivery.
Sigma-Aldrich
1-Adamantanecarbonyl chloride, 95%