MilliporeSigma
  • Ontogenesis of evolved changes in respiratory physiology in deer mice native to high altitude.
Ontogenesis of evolved changes in respiratory physiology in deer mice native to high altitude.
The Journal of experimental biology (2020-02-15)
Catherine M Ivy, Mary A Greaves, Elizabeth D Sangster, Cayleih E Robertson, Chandrasekhar Natarajan, Jay F Storz, Grant B McClelland, Graham R Scott
ABSTRACT
High-altitude environments are cold and hypoxic, and many high-altitude natives have evolved changes in respiratory physiology that improve O2 uptake in hypoxia as adults. Altricial mammals undergo a dramatic metabolic transition from ectothermy to endothermy in early post-natal life, which may influence the ontogenetic development of respiratory traits at high altitude. We examined the developmental changes in respiratory and haematological traits in deer mice (Peromyscus maniculatus) native to high altitude, comparing the respiratory responses to progressive hypoxia between highland and lowland deer mice. Among adults, highlanders exhibited higher total ventilation and a more effective breathing pattern (relatively deeper tidal volumes), for mice that were caught and tested at their native altitudes and those lab-raised in normoxia. Lab-raised progeny of each population were also tested at post-natal day (P)7, 14, 21 and 30. Highlanders developed an enhanced hypoxic ventilatory response by P21, concurrent with the full maturation of the carotid bodies, and their more effective breathing pattern arose by P14; these ages correspond to critical benchmarks in the full development of homeothermy in highlanders. However, highlanders exhibited developmental delays in ventilatory sensitivity to hypoxia, hyperplasia of type I cells in the carotid body and increases in blood haemoglobin content compared with lowland mice. Nevertheless, highlanders maintained consistently higher arterial O2 saturation in hypoxia across development, in association with increases in blood-O2 affinity that were apparent from birth. We conclude that evolved changes in respiratory physiology in high-altitude deer mice become expressed in association with the post-natal development of endothermy.
Sigma-Aldrich
Monoclonal Anti-Growth Associated Protein-43 antibody produced in mouse, clone GAP-7B10, ascites fluid
Sigma-Aldrich
Anti-Neurofilament 70 kDa Antibody, clone DA2, culture supernatant, clone DA2, Chemicon®
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit