MilliporeSigma
  • DNA methylation changes during preimplantation development reveal inter-species differences and reprogramming events at imprinted genes.
DNA methylation changes during preimplantation development reveal inter-species differences and reprogramming events at imprinted genes.
Clinical epigenetics (2020-05-13)
Elena Ivanova, Sebastian Canovas, Soledad Garcia-Martínez, Raquel Romar, Jordana S Lopes, Dimitrios Rizos, Maria J Sanchez-Calabuig, Felix Krueger, Simon Andrews, Fernando Perez-Sanz, Gavin Kelsey, Pilar Coy
ABSTRACT
Preimplantation embryos experience profound resetting of epigenetic information inherited from the gametes. Genome-wide analysis at single-base resolution has shown similarities but also species differences between human and mouse preimplantation embryos in DNA methylation patterns and reprogramming. Here, we have extended such analysis to two key livestock species, the pig and the cow. We generated genome-wide DNA methylation and whole-transcriptome datasets from gametes to blastocysts in both species. In oocytes from both species, a distinctive bimodal methylation landscape is present, with hypermethylated domains prevalent over hypomethylated domains, similar to human, while in the mouse the proportions are reversed.An oocyte-like pattern of methylation persists in the cleavage stages, albeit with some reduction in methylation level, persisting to blastocysts in cow, while pig blastocysts have a highly hypomethylated landscape. In the pig, there was evidence of transient de novo methylation at the 8-16 cell stages of domains unmethylated in oocytes, revealing a complex dynamic of methylation reprogramming. The methylation datasets were used to identify germline differentially methylated regions (gDMRs) of known imprinted genes and for the basis of detection of novel imprinted loci. Strikingly in the pig, we detected a consistent reduction in gDMR methylation at the 8-16 cell stages, followed by recovery to the blastocyst stage, suggesting an active period of imprint stabilization in preimplantation embryos. Transcriptome analysis revealed absence of expression in oocytes of both species of ZFP57, a key factor in the mouse for gDMR methylation maintenance, but presence of the alternative imprint regulator ZNF445. In conclusion, our study reveals species differences in DNA methylation reprogramming and suggests that porcine or bovine models may be closer to human in key aspects than in the mouse model.
Sigma-Aldrich
Imprint® DNA Modification Kit, For bisulfite DNA conversion & purification