MilliporeSigma
  • Indoor spraying with chlorfenapyr (a pyrrole insecticide) provides residual control of pyrethroid-resistant malaria vectors in southern Benin.
Indoor spraying with chlorfenapyr (a pyrrole insecticide) provides residual control of pyrethroid-resistant malaria vectors in southern Benin.
Malaria journal (2020-07-15)
Corine Ngufor, Augustin Fongnikin, Neil Hobbs, Martial Gbegbo, Laurette Kiki, Abibath Odjo, Martin Akogbeto, Mark Rowland
ABSTRACT
New classes of insecticides with novel modes of action, which can provide effective and prolonged control of insecticide-resistant malaria vector populations, are urgently needed for indoor residual spraying. Such insecticides can be included in a rotation plan to manage and prevent further development of resistance in mosquito vectors of malaria. Chlorfenapyr, a novel pyrrole insecticide with a unique mode of action, is being developed as a long-lasting IRS formulation. The efficacy of several formulations of chlorfenapyr alone and as mixtures with alpha-cypermethrin were evaluated in an experimental hut trial against wild pyrethroid-resistant Anopheles gambiae sensu lato in Cové, Benin, in an attempt to identify the most effective and long-lasting formulations for IRS. The trial lasted 12 months. A comparison was made with alpha-cypermethrin and bendiocarb formulations. CDC bottle bioassays were performed to investigate cross-resistance to chlorfenapyr in the local vector population. Mortality rates in World Health Organization (WHO) cylinder bioassays were < 5% with pyrethroids due to high levels of pyrethroid resistance, but > 95% with bendiocarb thus confirming susceptibility to carbamates in the vector population. CDC bottle bioassays showed no cross-resistance between pyrethroids and chlorfenapyr. Overall mortality of free-flying mosquitoes entering the experimental huts over the 12-month trial was 4% with alpha-cypermethrin and 12% with bendiocarb. The chlorfenapyr solo-formulations induced significantly higher levels of mortality (38-46%) compared to the bendiocarb (12% P < 0.001) and to the mixture formulations (18-22%, P < 0.05). The original Sylando 240SC formulation of chlorfenapyr was more efficacious than all other novel chlorfenapyr formulations tested. Bendiocarb induced > 80% mortality in the first month, but this declined sharply to < 20% by the third month while the mortality rates achieved with the chlorfenapyr formulations (38-46%) were persistent lasting 7-10 months. The mixtures induced significantly lower percentage mortality than chlorfenapyr-solo formulations. Wall cone bioassays only showed mortality rates that were consistent with chlorfenapyr IRS treated huts when the exposure time was increased to 2 h. Indoor residual spraying with chlorfenapyr (Sylando® 240SC) provides moderate but prolonged control of pyrethroid-resistant malaria vectors compared to pyrethroid and bendiocarb IRS. Wall cone bioassays on chlorfenapyr-treated walls required longer exposure times of 2 h than the customary 30 min indicating that WHO guidelines on residual cone bioassays need to be more insecticide-specific.
Product Description
Supelco
Bendiocarb, PESTANAL®, analytical standard