MilliporeSigma
  • Unbiased Proteomic Profiling Uncovers a Targetable GNAS/PKA/PP2A Axis in Small Cell Lung Cancer Stem Cells.
Unbiased Proteomic Profiling Uncovers a Targetable GNAS/PKA/PP2A Axis in Small Cell Lung Cancer Stem Cells.
Cancer cell (2020-06-13)
Garry L Coles, Sandra Cristea, James T Webber, Rebecca S Levin, Steven M Moss, Andy He, Jaya Sangodkar, Yeonjoo C Hwang, Julia Arand, Alexandros P Drainas, Nancie A Mooney, Janos Demeter, Jessica N Spradlin, Brandon Mauch, Vicky Le, Yan Ting Shue, Julie H Ko, Myung Chang Lee, Christina Kong, Daniel K Nomura, Michael Ohlmeyer, Danielle L Swaney, Nevan J Krogan, Peter K Jackson, Goutham Narla, John D Gordan, Kevan M Shokat, Julien Sage
ABSTRACT
Using unbiased kinase profiling, we identified protein kinase A (PKA) as an active kinase in small cell lung cancer (SCLC). Inhibition of PKA activity genetically, or pharmacologically by activation of the PP2A phosphatase, suppresses SCLC expansion in culture and in vivo. Conversely, GNAS (G-protein α subunit), a PKA activator that is genetically activated in a small subset of human SCLC, promotes SCLC development. Phosphoproteomic analyses identified many PKA substrates and mechanisms of action. In particular, PKA activity is required for the propagation of SCLC stem cells in transplantation studies. Broad proteomic analysis of recalcitrant cancers has the potential to uncover targetable signaling networks, such as the GNAS/PKA/PP2A axis in SCLC.
Sigma-Aldrich
ApopTag Fluorescein In Situ Apoptosis Detection Kit, The ApopTag Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a Fluorescein reporter molecule.
Sigma-Aldrich
Anti-Neural Cell Adhesion Molecule Antibody, Chemicon®, from rabbit