MilliporeSigma
  • Spherical mesoporous covalent organic framework as a solid-phase extraction adsorbent for the ultrasensitive determination of sulfonamides in food and water samples by liquid chromatography-tandem mass spectrometry.
Spherical mesoporous covalent organic framework as a solid-phase extraction adsorbent for the ultrasensitive determination of sulfonamides in food and water samples by liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2020-07-28)
Lian Wen, Lu Liu, Xia Wang, Ming-Lin Wang, Jin-Ming Lin, Ru-Song Zhao
ABSTRACT
Efficient extraction of polar sulfonamides antibiotics from aqueous samples and food is very challenging, because they are hydrophilic, their concentration is very low, and the matrix is complex. Covalent organic frameworks (COFs), a novel porous organic material, have attracted great attention. In this work, the spherical triphenylbenzene-dimethoxyterephthaldehyde-COFs (TPB-DMTP-COFs) were synthesized by a simple room temperature method, and due to their attractive properties, such as high outstanding acid-base stability, large specific surface area, low skeletal density, inherent porosity and high crystallinity, so TPB-DMTP-COFs as ideal solid phase extraction adsorbents showed excellent adsorption performance for trace polar sulfonamides in food and water. TPB-DMTP-COFs were characterized by scanning electron microscopy, Fourier-transform infrared spectroscopy, powder X-ray diffraction, and so on. The important parameters were optimized to improve the extraction efficiency of TPB-DMTP-COFs toward sulfonamides. Analysis of sulfonamides was performed by liquid chromatography-tandem mass spectrometry. The developed method based on TPB-DMTP-COFs material achieved low limits of detection (0.5-1.0 ng L-1), wide linearity (5-1000 ng L-1), and good repeatability (2.5%-8.7%). The possible extraction mechanism was also discussed. Finally, the method was successfully applied to the enrichment and detection of sulfonamides in environmental water samples and food samples. The present study indicated that TPB-DMTP-COFs had splendid prospects in highly sensitive analysis of other pollutants in complex matrix.
Sigma-Aldrich
2,5-Dimethoxybenzene-1,4-dicarboxaldehyde, 97%