MilliporeSigma
  • Reproduction in the Eared Dove: An exception to the classic model of seasonal reproduction in birds?
Reproduction in the Eared Dove: An exception to the classic model of seasonal reproduction in birds?
Zoology (Jena, Germany) (2020-04-07)
Ludmila Maldonado, Tomas C Tempesti, Gustavo M Somoza, Susana I Peluc, Diego J Valdez
ABSTRACT
In avian species living at high altitudes and latitudes, reproductive events are largely controlled by photoperiod, with changes being perceived mainly through encephalic photoreceptors located in the hypothalamus. It is known that during long day periods (reproductive periods), the information transmitted by brain photoreceptors triggers the production of thyroid hormones that regulate GnRH secretion, inducing secretion of pituitary gonadotropins. As a result, gonads develop and grow and the production of gonadal sex hormones, testosterone and estradiol increases (classic gonadal cycle). During short day periods (non-reproductive periods) on the other hand, the gonads regress, and plasma gonadal steroid levels are low. By means of this mechanism, birds synchronize their physiology and reproductive behaviors with seasonal changes in the environment. However, it appears that not all avian species comply with this general reproductive pattern. For example, the Eared Dove (Zenaida auriculata), a South American opportunistic breeding columbiform, has been reported to successfully reproduce throughout the year, making it an interesting avian system for studying the endocrine basis of avian reproduction. In view of a clear lack of seasonal variability in testicular weight and size (the classic gonadal regression/recrudescence cycle) in the male Eared Dove, we examined whether their reproductive aseasonality could be the result of being in a continuous state of reproductive preparedness. Our results show that despite the absence of a marked gonadal cycle in terms of gonadal volume, plasma testosterone levels in males were minimal during autumn-winter, reaching maximum values during spring-summer. This indicates that male gonad function is not seasonal in terms of spermatogenesis but that circulating testosterone levels are correlated with photoperiod, demonstrating an exception to the classic model of reproduction in birds.