MilliporeSigma
A short de novo synthesis of nucleoside analogs.
Science (New York, N.Y.) (2020-08-09)
Michael Meanwell, Steven M Silverman, Johannes Lehmann, Bharanishashank Adluri, Yang Wang, Ryan Cohen, Louis-Charles Campeau, Robert Britton
ABSTRACT
Nucleoside analogs are commonly used in the treatment of cancer and viral infections. Their syntheses benefit from decades of research but are often protracted, unamenable to diversification, and reliant on a limited pool of chiral carbohydrate starting materials. We present a process for rapidly constructing nucleoside analogs from simple achiral materials. Using only proline catalysis, heteroaryl-substituted acetaldehydes are fluorinated and then directly engaged in enantioselective aldol reactions in a one-pot reaction. A subsequent intramolecular fluoride displacement reaction provides a functionalized nucleoside analog. The versatility of this process is highlighted in multigram syntheses of d- or l-nucleoside analogs, locked nucleic acids, iminonucleosides, and C2'- and C4'-modified nucleoside analogs. This de novo synthesis creates opportunities for the preparation of diversity libraries and will support efforts in both drug discovery and development.
Sigma-Aldrich
D-Proline, ReagentPlus®, ≥99%