MilliporeSigma
  • A circular RNA map for human induced pluripotent stem cells of foetal origin.
A circular RNA map for human induced pluripotent stem cells of foetal origin.
EBioMedicine (2020-06-24)
Mario Barilani, Alessandro Cherubini, Valeria Peli, Francesca Polveraccio, Valentina Bollati, Federica Guffanti, Alessandro Del Gobbo, Cristiana Lavazza, Silvia Giovanelli, Nicola Elvassore, Lorenza Lazzari
ABSTRACT
Adult skin fibroblasts represent the most common starting cell type used to generate human induced pluripotent stem cells (F-hiPSC) for clinical studies. Yet, a foetal source would offer unique advantages, primarily the absence of accumulated somatic mutations. Herein, we generated hiPSC from cord blood multipotent mesenchymal stromal cells (MSC-hiPSC) and compared them with F-hiPSC. Assessment of the full activation of the pluripotency gene regulatory network (PGRN) focused on circular RNA (circRNA), recently proposed to participate in the control of pluripotency. Reprogramming was achieved by a footprint-free strategy. Self-renewal and pluripotency of cord blood MSC-hiPSC were investigated in vitro and in vivo, compared to parental MSC, to embryonic stem cells and to F-hiPSC. High-throughput array-based approaches and bioinformatics analyses were applied to address the PGRN. Cord blood MSC-hiPSC successfully acquired a complete pluripotent identity. Functional comparison with F-hiPSC showed no differences in terms of i) generation of mesenchymal-like derivatives, ii) their subsequent adipogenic, osteogenic and chondrogenic commitment, and iii) their hematopoietic support ability. At the transcriptional level, specific subsets of mRNA, miRNA and circRNA (n = 4,429) were evidenced, casting a further layer of complexity on the PGRN regulatory crosstalk. A circRNA map of transcripts associated to naïve and primed pluripotency is provided for hiPSC of clinical-grade foetal origin, offering insights on still unreported regulatory circuits of the PGRN to consider for the optimization and development of efficient differentiation protocols for clinical translation. This research was funded by Ricerca Corrente 2012-2018 by the Italian Ministry of Health.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Greiner CELLSTAR® 96 well plates, V-bottom clear wells