MilliporeSigma
  • Expression of Insulin-Like Growth Factor 1 (IGF-1) and Epidermal Growth Factor (EGF) Receptors and the Effect of IGF-1 and EGF on Androgen and Estrogen Release in the Myometrium of Pigs-In Vitro Study.
Expression of Insulin-Like Growth Factor 1 (IGF-1) and Epidermal Growth Factor (EGF) Receptors and the Effect of IGF-1 and EGF on Androgen and Estrogen Release in the Myometrium of Pigs-In Vitro Study.
Animals : an open access journal from MDPI (2020-05-30)
Ewa Monika Waszkiewicz, Wiktoria Kozlowska, Agata Zmijewska, Anita Franczak
ABSTRACT
Porcine myometrium possesses steroidogenic activity but its regulation is not well understood. It was hypothesized that the regulators of myometrial steroidogenesis are insulin-like growth factor 1 (IGF-1) and epidermal growth factor (EGF), which were found to modulate the steroidogenic activity of the endometrium and embryos. Myometrial slices were collected from gravid and nongravid pigs on days 10 or 11, 12 or 13 and 15 or 16 and studied for: (1) the relative abundance of IGF-1R and EGFR mRNA transcripts and proteins, to determine myometrial readiness to response growth factors treatment and (2) the effect of IGF-1 or EGF on the myometrial release of androstenedione (A4), testosterone (T), estrone (E1) and estradiol-17β (E2). The results showed that the relative expression and abundance of IGF-1R and EGFR in the myometrium were altered regarding the female reproductive status. During the estrous cycle, EGF increased myometrial release of A4 on days 12-13 and E2 on days 15-16. In gravid pigs (days 15-16), IGF-1 and EGF increased the E1 release. In conclusion: (1) porcine myometrium possesses the potential to respond to IGF-1 and EGF treatment, (2) EGF significantly increases myometrial A4 and E2 release in cyclic pigs, while IGF-1 and EGF increase the E1 release in gravid pigs.
Sigma-Aldrich
Fluoroshield with DAPI, liquid
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
GenElute Gel Extraction Kit, sufficient for 70 purifications