MilliporeSigma
  • Human Mesenchymal Stem Cell Secretome Exhibits a Neuroprotective Effect over In Vitro Retinal Photoreceptor Degeneration.
Human Mesenchymal Stem Cell Secretome Exhibits a Neuroprotective Effect over In Vitro Retinal Photoreceptor Degeneration.
Molecular therapy. Methods & clinical development (2020-06-10)
Ricardo Usategui-Martín, Kevin Puertas-Neyra, María-Teresa García-Gutiérrez, Manuel Fuentes, José Carlos Pastor, Ivan Fernandez-Bueno
ABSTRACT
Retinal photoreceptor degeneration occurs frequently in several neurodegenerative retinal diseases such as age-related macular degeneration, retinitis pigmentosa, or genetic retinal diseases related to the photoreceptors. Despite the impact on daily life and the social and economic consequences, there is no cure for these diseases. Considering this, cell-based therapy may be an optimal therapeutic option. This study evaluated the neuroprotective in vitro potential of a secretome of human bone marrow mesenchymal stem cells (MSCs) for retinal photoreceptors in vitro. We analyzed the photoreceptor morphologic changes and the paracrine factors secreted by human bone marrow MSCs in a physically separated co-culture with degenerated neuroretinas, using organotypic neuroretinal cultures. The results showed that the secretome of human bone marrow MSCs had a neuroprotective effect over the neuroretinal general organization and neuropreserved the photoreceptors from degeneration probably by secretion of neuroprotective proteins. The study of the expression of 1,000 proteins showed increased paracrine factors secreted by MSCs that could be crucial in the neuroprotective effect of the stem cell secretome over in vitro retinal degeneration. The current results reinforce the hypothesis that the paracrine effect of the human bone marrow MSCs may slow photoreceptor neurodegeneration and be a therapeutic option in retinal photoreceptor degenerative diseases.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Magnetic Stir Bar, for use with For 3.0-5.0mL vessels, pkg of 6 ea, PTFE
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Human Mesenchymal Stem Cell Characterization Kit, The Human Mesenchymal Stem Cell Characterization Kit contains a panel of positive & negative selection markers for the characterization of the mesenchymal stem cell population in human samples.