MilliporeSigma
  • Simultaneous separation and determination of 20 potential adulterant antigout and antiosteoporosis pharmaceutical compounds in herbal food products using LC with electrospray ionization MS/MS and LC with quadrupole-time-of-flight MS.
Simultaneous separation and determination of 20 potential adulterant antigout and antiosteoporosis pharmaceutical compounds in herbal food products using LC with electrospray ionization MS/MS and LC with quadrupole-time-of-flight MS.
Journal of separation science (2020-04-17)
Nam Sook Kim, Sun Hee Moon, Hwan Seong Choi, Ji Hyun Lee, Seongsoo Park, Hoil Kang
ABSTRACT
An analytical method for the simultaneous and reliable determination of 20 antigout and antiosteoporosis pharmaceutical compounds in adulterated health food products was developed using liquid chromatography with electrospray ionization tandem mass spectrometry and liquid chromatography with quadrupole-time-of-flight mass spectrometry. The method was validated through the determination of specificity, linearity, limit of detection, and limit of quantification, method detection limit, method quantitation limit, precision, accuracy, recovery, and stability. The matrix effect was also determined. The validation results of the developed method are as follows: for solid and liquid blank samples, limits of detection ranged from 0.05 to 5.00 ng/mL and limits of quantification ranged from 0.15 to 15.00 ng/mL. Linearity was acceptable, and the correlation coefficients (R2 ) were ≥0.99 for all target compounds. Both intra and interday precision were less than 9.16% RSD, and accuracies ranged from 95.31 to 116.68%. Mean recoveries for different types of dietary supplements classified as powders, liquids, tablets, and capsules were found to be 80.81 to 117.62% with less than 15.00% relative standard deviation. The stability of the standard mixture solution was less than 11.72% relative standard deviation after 48 h. By the proposed method, the presence of dexamethasone was determined in seized herbal food products at concentrations that ranged from 126 to 215 µg/g.
Sigma-Aldrich
Lasofoxifene tartrate, ≥98% (HPLC)
Sigma-Aldrich
2-Ethoxybenzamide, 97%