MilliporeSigma
  • Inefficient thermogenic mitochondrial respiration due to futile proton leak in a mouse model of fragile X syndrome.
Inefficient thermogenic mitochondrial respiration due to futile proton leak in a mouse model of fragile X syndrome.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-04-21)
Keren K Griffiths, Aili Wang, Lifei Wang, Matthew Tracey, Giulio Kleiner, Catarina M Quinzii, Linlin Sun, Guang Yang, Jose F Perez-Zoghbi, Pawel Licznerski, Mu Yang, Elizabeth A Jonas, Richard J Levy
ABSTRACT
Fragile X syndrome (FXS) is the leading known inherited intellectual disability and the most common genetic cause of autism. The full mutation results in transcriptional silencing of the Fmr1 gene and loss of fragile X mental retardation protein (FMRP) expression. Defects in neuroenergetic capacity are known to cause a variety of neurodevelopmental disorders. Thus, we explored the integrity of forebrain mitochondria in Fmr1 knockout mice during the peak of synaptogenesis. We found inefficient thermogenic respiration due to futile proton leak in Fmr1 KO mitochondria caused by coenzyme Q (CoQ) deficiency and an open cyclosporine-sensitive channel. Repletion of mitochondrial CoQ within the Fmr1 KO forebrain closed the channel, blocked the pathological proton leak, restored rates of protein synthesis during synaptogenesis, and normalized the key phenotypic features later in life. The findings demonstrate that FMRP deficiency results in inefficient oxidative phosphorylation during the neurodevelopment and suggest that dysfunctional mitochondria may contribute to the FXS phenotype.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Coenzyme Q10, ≥98% (HPLC)
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Sigma-Aldrich
Citrate Synthase Assay Kit, 1 kit sufficient for 100 reactions (using a 1 ml cuvette), 1 kit sufficient for 480 reactions (using 96 multiwell plates)
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse