MilliporeSigma
  • Acquired Resistance to Alectinib in ALK-Rearranged Lung Cancer due to ABCC11/MRP8 Overexpression in a Clinically Paired Resistance Model.
Acquired Resistance to Alectinib in ALK-Rearranged Lung Cancer due to ABCC11/MRP8 Overexpression in a Clinically Paired Resistance Model.
Molecular cancer therapeutics (2020-03-29)
Tomoko Funazo, Takahiro Tsuji, Hiroaki Ozasa, Koh Furugaki, Yasushi Yoshimura, Tetsuya Oguri, Hitomi Ajimizu, Yuto Yasuda, Takashi Nomizo, Yuichi Sakamori, Hironori Yoshida, Young Hak Kim, Toyohiro Hirai
ABSTRACT
Alectinib is used as a first-line treatment for anaplastic lymphoma kinase (ALK)-rearranged non-small cell lung cancer (NSCLC). Whereas other ALK inhibitors have been reported to be involved in resistance to ATP-binding cassette (ABC) transporters, no data are available regarding the association between resistance to alectinib and ABC-transporters. To investigate whether ABC-transporters contribute to alectinib resistance, ABC-transporter expression in alectinib-resistant cell lines derived from a patient with ALK-rearranged NSCLC and from H2228 lung cancer cells was evaluated and compared with that in each parent cell type. ATP-binding cassette subfamily C member 11 (ABCC11) expression was significantly increased in alectinib-resistant cell lines compared with that in alectinib-sensitive lines. ABCC11 inhibition increased sensitivity to alectinib in vitro ABCC11-overexpressing cells were established by transfection of an ABCC11 expression vector into H2228 cells, while control cells were established by transfecting H2228 cells with an empty vector. ABCC11-overexpressing cells exhibited decreased sensitivity to alectinib compared with that of control cells in vitro Moreover, the tumor growth rate following alectinib treatment was higher in ABCC11-overexpressing cells than that in control cells in vivo In addition, the intracellular alectinib concentration following exposure to 100 nmol/L alectinib was significantly lower in the ABCC11-overexpressing cell line compared with that in control cells. This is the first preclinical evidence showing that ABCC11 expression may be involved in acquired resistance to alectinib.