MilliporeSigma
  • Hemorheology and oxidative stress in patients with differentiated thyroid cancer following I-131 ablation/metastasis treatment.
Hemorheology and oxidative stress in patients with differentiated thyroid cancer following I-131 ablation/metastasis treatment.
Clinical hemorheology and microcirculation (2019-12-31)
Tarik Sengoz, Emine Kilic-Toprak, Olga Yaylali, Ozgen Kilic-Erkek, Yasin Ozdemir, Burak Oymak, Hande Senol, Dogangun Yuksel, Vural Kucukatay, Melek Bor-Kucukatay
ABSTRACT
Although radioiodine theraphy (RAIT) is thought to affect blood cells and oxidative stress, hemorheological alterations following dose-dependent RAIT remains unknown. The aim of this study was to determine the effects of RAIT on hemorheological and oxidative stress parameters in patients with differentiated thyroid cancers (DTC). Totally 31 DTC patients (mean age 46.32±11.15 years) and 26 healthy controls (mean age 50.50±6.22 years) were included. Venous blood samples were collected from each patient before and after treatment (7th day, 1th month and 6th month). Erythrocyte aggregation-deformability and oxidative stress parameters were determined. p < 0.05 was considered as statistically significant. Erythrocyte deformability of the patients determined at 16.87 and 30 Pascal were significantly lower than healthy individuals. Erythrocyte aggregation index (AI) of the patients was higher, whereas erythrocyte aggregation half-time (t½) was lower compared to control. Erythrocyte deformability values and AI were not significantly different from the pre- and post-radioiodine treatment groups. There was no statistically significant difference between the oxidative stress parameters before and after the treatment. Patients were in a worse hemorheological condition compared to healthy individuals. After RAIT, RBC deformability and aggregation were not affected and no significant change in oxidative stress parameters was detected.
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000