MilliporeSigma
  • Melatonin-related mitochondrial respiration responses are associated with growth promotion and cold tolerance in plants.
Melatonin-related mitochondrial respiration responses are associated with growth promotion and cold tolerance in plants.
Cryobiology (2019-11-24)
Hulya Turk, Mucip Genisel
ABSTRACT
Melatonin has the ability to improve plant growth and strengthened plant tolerance to environmental stresses; however, the effects of melatonin on mitochondrial respiration in plants and the underlying biochemical and molecular mechanisms are still unclear. The objective of the study is to determine possible effects of melatonin on mitochondrial respiration and energy efficiency in maize leaves grown under optimum temperature and cold stress and to reveal the relationship between melatonin-induced possible alterations in mitochondrial respiration and cold tolerance. Melatonin and cold stress, alone and in combination, caused significant increases in activities and gene expressions of pyruvate dehydrogenase, citrate synthase, and malate dehydrogenase, indicating an acceleration in the rate of tricarboxylic acid cycle. Total mitochondrial respiration rate, cytochrome pathway rate, and alternative respiration rate were increased by the application of melatonin and/or cold stress. Similarly, gene expression and protein levels of cytochrome oxidase and alternative oxidase were also enhanced by melatonin and/or cold stress. The highest values for all these parameters were obtained from the seedlings treated with the combined application of melatonin and cold stress. The activity and gene expression of ATP synthase and ATP concentration were augmented by melatonin under control and cold stress. On the other hand, cold stress reduced markedly plant growth parameters, including root length, plant height, leaf surface area, and chlorophyll content and increased the content of reactive oxygen species (ROS), including superoxide anion and hydrogen peroxide and oxidative damage, including malondialdehyde content and electrolyte leakage level; however, melatonin significantly promoted the plant growth parameters and reduced ROS content and oxidative damage under control and cold stress. These data revealed that melatonin-induced growth promotion and cold tolerance in maize is associated with its modulating effect on mitochondrial respiration.
Sigma-Aldrich
Citrate Synthase Assay Kit, 1 kit sufficient for 100 reactions (using a 1 ml cuvette), 1 kit sufficient for 480 reactions (using 96 multiwell plates)