MilliporeSigma
  • Regulation of BMP4/Dpp retrotranslocation and signaling by deglycosylation.
Regulation of BMP4/Dpp retrotranslocation and signaling by deglycosylation.
eLife (2020-07-29)
Antonio Galeone, Joshua M Adams, Shinya Matsuda, Maximiliano F Presa, Ashutosh Pandey, Seung Yeop Han, Yuriko Tachida, Hiroto Hirayama, Thomas Vaccari, Tadashi Suzuki, Cathleen M Lutz, Markus Affolter, Aamir Zuberi, Hamed Jafar-Nejad
ABSTRACT
During endoplasmic reticulum-associated degradation (ERAD), the cytoplasmic enzyme N-glycanase 1 (NGLY1) is proposed to remove N-glycans from misfolded N-glycoproteins after their retrotranslocation from the ER to the cytosol. We previously reported that NGLY1 regulates Drosophila BMP signaling in a tissue-specific manner (Galeone et al., 2017). Here, we establish the Drosophila Dpp and its mouse ortholog BMP4 as biologically relevant targets of NGLY1 and find, unexpectedly, that NGLY1-mediated deglycosylation of misfolded BMP4 is required for its retrotranslocation. Accumulation of misfolded BMP4 in the ER results in ER stress and prompts the ER recruitment of NGLY1. The ER-associated NGLY1 then deglycosylates misfolded BMP4 molecules to promote their retrotranslocation and proteasomal degradation, thereby allowing properly-folded BMP4 molecules to proceed through the secretory pathway and activate signaling in other cells. Our study redefines the role of NGLY1 during ERAD and suggests that impaired BMP4 signaling might underlie some of the NGLY1 deficiency patient phenotypes.
Sigma-Aldrich
Anti-Green Fluorescent Protein (GFP) antibody, Mouse monoclonal, clone GSN149, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NGLY1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-HA tag antibody produced in mouse, clone GT4810, affinity isolated antibody
Sigma-Aldrich
Anti-Ubiquitinylated proteins Antibody, clone FK1, clone FK1, Upstate®, from mouse