MilliporeSigma
  • Elevated concentration of cerebrospinal fluid tissue transglutaminase in Parkinson's disease indicating apoptosis.
Elevated concentration of cerebrospinal fluid tissue transglutaminase in Parkinson's disease indicating apoptosis.
Movement disorders : official journal of the Movement Disorder Society (2004-09-16)
Istvan Vermes, Ernst N H Jansen Steur, Gustav F Jirikowski, Clemens Haanen
ABSTRACT
Tissue transglutaminase (tTG) is activated during the apoptotic cell death cascade and plays a key role in the formation of apoptotic bodies. We found significant elevation of tTG concentration in the cerebrospinal fluid (CSF) of 54 patients with Parkinson's disease (PD) compared with that measured in 34 control subjects, thus showing increased neural cell apoptosis in patients with PD.
Sigma-Aldrich
Microbial Transglutaminase, lyophilized powder, ≥12 units/mg protein