MilliporeSigma
  • Highly Reproducible Perovskite Solar Cells with Average Efficiency of 18.3% and Best Efficiency of 19.7% Fabricated via Lewis Base Adduct of Lead(II) Iodide.
Highly Reproducible Perovskite Solar Cells with Average Efficiency of 18.3% and Best Efficiency of 19.7% Fabricated via Lewis Base Adduct of Lead(II) Iodide.
Journal of the American Chemical Society (2015-07-01)
Namyoung Ahn, Dae-Yong Son, In-Hyuk Jang, Seong Min Kang, Mansoo Choi, Nam-Gyu Park
ABSTRACT
High efficiency perovskite solar cells were fabricated reproducibly via Lewis base adduct of lead(II) iodide. PbI2 was dissolved in N,N-dimethyformamide with equimolar N,N-dimethyl sulfoxide (DMSO) and CH3NH3I. Stretching vibration of S═O appeared at 1045 cm(-1) for bare DMSO, which was shifted to 1020 and 1015 cm(-1) upon reacting DMSO with PbI2 and PbI2 + CH3NH3I, respectively, indicative of forming the adduct of PbI2·DMSO and CH3NH3I·PbI2·DMSO due to interaction between Lewis base DMSO and/or iodide (I(-)) and Lewis acid PbI2. Spin-coating of a DMF solution containing PbI2, CH3NH3I, and DMSO (1:1:1 mol %) formed a transparent adduct film, which was converted to a dark brown film upon heating at low temperature of 65 °C for 1 min due to removal of the volatile DMSO from the adduct. The adduct-induced CH3NH3PbI3 exhibited high charge extraction characteristics with hole mobility as high as 3.9 × 10(-3) cm(2)/(V s) and slow recombination rate. Average power conversion efficiency (PCE) of 18.3% was achieved from 41 cells and the best PCE of 19.7% was attained via adduct approach.
Sigma-Aldrich
Lithium bis(trifluoromethanesulfonyl)imide, anhydrous, 99.99% trace metals basis