MilliporeSigma
  • Composition, antioxidant, antimicrobial and anti-wood-decay fungal activities of the twig essential oil of Taiwania cryptomerioides from Taiwan.
Composition, antioxidant, antimicrobial and anti-wood-decay fungal activities of the twig essential oil of Taiwania cryptomerioides from Taiwan.
Natural product communications (2012-04-06)
Chen-Lung Ho, Su-Sing Yang, Tsong-Min Chang, Yu-Chang Su
ABSTRACT
This study investigated the chemical composition, antioxidant, antimicrobial and anti-wood-decay fungal activities of the essential oil isolated from the twigs of Taiwania cryptomerioides from Taiwan. The essential oil was isolated using hydrodistillation in a Clevenger-type apparatus, and characterized by GC-FID and GC-MS. A total of 35 compounds were identified, representing 100% of the oil. The main components identified were alpha-cadinol (45.9%), ferruginol (18.9%) and beta-eudesmol (10.8%). The antioxidant activity of the oil was tested by the DPPH (2,2-diphenyl-1-picrylhydrazyl) free radical scavenging capability test. The results showed an IC50 of 90.8 +/- 0.2 microg/mL. The active source compound was ferruginol. The antimicrobial activity of the oil was tested by the disc diffusion and micro-broth dilution methods against ten microbial species. The oil exhibited strong growth suppression against Gram-positive bacteria and yeast with inhibition zones of 45-52 mm and MIC values of 31.25-62.5 microg/mL, respectively. The anti-wood-decay fungal activity of the oil was also evaluated. The oil demonstrated excellent activity against four wood-decay-fungal species. For the antimicrobial and anti-wood-decay fungal activities of the oil, the active source compounds were determined to be alpha-cadinol, beta-eudesmol and ferruginol.
Sigma-Aldrich
β-Eudesmol, ≥90% (GC)