MilliporeSigma
  • Resolution of R-loops by INO80 promotes DNA replication and maintains cancer cell proliferation and viability.
Resolution of R-loops by INO80 promotes DNA replication and maintains cancer cell proliferation and viability.
Nature communications (2020-09-12)
Lisa Prendergast, Urszula L McClurg, Rossitsa Hristova, Rolando Berlinguer-Palmini, Sarah Greener, Katie Veitch, Inmaculada Hernandez, Philippe Pasero, Daniel Rico, Jonathan M G Higgins, Anastas Gospodinov, Manolis Papamichos-Chronakis
ABSTRACT
Collisions between the DNA replication machinery and co-transcriptional R-loops can impede DNA synthesis and are a major source of genomic instability in cancer cells. How cancer cells deal with R-loops to proliferate is poorly understood. Here we show that the ATP-dependent chromatin remodelling INO80 complex promotes resolution of R-loops to prevent replication-associated DNA damage in cancer cells. Depletion of INO80 in prostate cancer PC3 cells leads to increased R-loops. Overexpression of the RNA:DNA endonuclease RNAse H1 rescues the DNA synthesis defects and suppresses DNA damage caused by INO80 depletion. R-loops co-localize with and promote recruitment of INO80 to chromatin. Artificial tethering of INO80 to a LacO locus enabled turnover of R-loops in cis. Finally, counteracting R-loops by INO80 promotes proliferation and averts DNA damage-induced death in cancer cells. Our work suggests that INO80-dependent resolution of R-loops promotes DNA replication in the presence of transcription, thus enabling unlimited proliferation in cancers.
Sigma-Aldrich
6-Hydroxy-DL-DOPA, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Histone H3 Antibody, 0.5 mg/mL, Upstate®
Sigma-Aldrich
Anti-DNA Antibody, single stranded, clone 16-19, clone 16-19, Chemicon®, from mouse