MilliporeSigma
  • Important role of interleukin-3 in the early phase of collagen-induced arthritis.
Important role of interleukin-3 in the early phase of collagen-induced arthritis.
Arthritis and rheumatism (2009-05-01)
Hilke Brühl, Josef Cihak, Marianne Niedermeier, Andrea Denzel, Manuel Rodriguez Gomez, Yvonne Talke, Nicole Goebel, Jirí Plachý, Manfred Stangassinger, Matthias Mack
ABSTRACT
Activation of basophils contributes to memory immune responses and results in exacerbation of collagen-induced arthritis (CIA). We undertook the present study to analyze the production and biologic effects of interleukin-3 (IL-3), a strong activator of basophils, in CIA. Arthritis was induced by immunization with type II collagen. Mice were treated with blocking monoclonal antibodies against IL-3 or with recombinant IL-3. Clinical scoring, histologic analysis, fluorescence-activated cell sorter analysis, enzyme-linked immunosorbent assay, and cell culturing were performed to assess disease activity and IL-3 production. IL-3 was produced in large quantities by collagen-specific CD4+ T cells in the spleen and was present in the synovial tissue during onset of arthritis, but was down-regulated in paws with severe inflammation. Blockade of IL-3 during the time of arthritis onset resulted in profound improvement of the disease, with reductions in synovial leukocyte and cytokine levels, peripheral blood basophil levels, and anticollagen antibody titers. Blockade of IL-3 during the late phase of arthritis had no beneficial effect. Administration of recombinant IL-3 during onset of arthritis induced a marked exacerbation of the disease, with increased peripheral blood basophil and plasma IL-6 levels and increased titers of anticollagen antibody. In studies of the regulation of IL-3 expression in CD4+ T cells, IL-6 and IL-4 suppressed the release of IL-3 by activated CD4+ T cells, whereas lipopolysaccharide and CpG DNA up-regulated IL-3 secretion in activated CD4+ T cells by acting on costimulatory cells. Taken together, the present results demonstrate for the first time that IL-3 has an important role in the early phase of CIA.
Sigma-Aldrich
Monoclonal Anti-SQSTM1 antibody produced in mouse, clone 2C11, purified immunoglobulin, buffered aqueous solution