MilliporeSigma
  • Fusion Stage of HIV-1 Entry Depends on Virus-Induced Cell Surface Exposure of Phosphatidylserine.
Fusion Stage of HIV-1 Entry Depends on Virus-Induced Cell Surface Exposure of Phosphatidylserine.
Cell host & microbe (2017-07-14)
Elena Zaitseva, Eugene Zaitsev, Kamran Melikov, Anush Arakelyan, Mariana Marin, Rafael Villasmil, Leonid B Margolis, Gregory B Melikyan, Leonid V Chernomordik
ABSTRACT
HIV-1 entry into host cells starts with interactions between the viral envelope glycoprotein (Env) and cellular CD4 receptors and coreceptors. Previous work has suggested that efficient HIV entry also depends on intracellular signaling, but this remains controversial. Here we report that formation of the pre-fusion Env-CD4-coreceptor complexes triggers non-apoptotic cell surface exposure of the membrane lipid phosphatidylserine (PS). HIV-1-induced PS redistribution depends on Ca2+ signaling triggered by Env-coreceptor interactions and involves the lipid scramblase TMEM16F. Externalized PS strongly promotes Env-mediated membrane fusion and HIV-1 infection. Blocking externalized PS or suppressing TMEM16F inhibited Env-mediated fusion. Exogenously added PS promoted fusion, with fusion dependence on PS being especially strong for cells with low surface density of coreceptors. These findings suggest that cell-surface PS acts as an important cofactor that promotes the fusogenic restructuring of pre-fusion complexes and likely focuses the infection on cells conducive to PS signaling.
Product Description
Avanti
16:0-06:0 NBD PS, Avanti Research - A Croda Brand 810192P, powder
Sigma-Aldrich
Calcium-activated Chloride Channel Inhibitor, CaCCinh-A01, The Calcium-activated Chloride Channel Inhibitor, CaCCinh-A01 controls the biological activity of Calcium-activated Chloride Channel.