MilliporeSigma
  • Further intracellular proteins and signaling pathways regulated by angiotensin-(1-7) in human endothelial cells.
Further intracellular proteins and signaling pathways regulated by angiotensin-(1-7) in human endothelial cells.
Data in brief (2016-12-27)
Christian Meinert, Franziska Kohse, Ilka Böhme, Florian Gembardt, Anja Tetzner, Thomas Wieland, Barry Greenberg, Thomas Walther
ABSTRACT
In 2016, Meinert et al. (doi: 10.1016/j.jprot.2015.09.020) published the first 25 proteins in a protein array regulated in Human Umbilical Vein Endothelial Cells (HUVEC) by the heptapeptide angiotensin (Ang)-(1-7) and the first 10 intracellular signaling cascades at different time points. This supporting data article shows further proteins and pathways stimulated by Ang-(1-7) in human endothelial cells at time points of 1 h, 3 h, 6 h, and 9 h. HUVECs were stimulated with Ang-(1-7), and regulated proteins were identified via antibody microarray. Bioinformatics software IPA was used for association of regulated proteins to metabolic pathways.