MilliporeSigma
  • Apoptotic sphingolipid signaling by ceramides in lung endothelial cells.
Apoptotic sphingolipid signaling by ceramides in lung endothelial cells.
American journal of respiratory cell and molecular biology (2008-01-15)
Terry R Medler, Daniela N Petrusca, Patty J Lee, Walter C Hubbard, Evgeny V Berdyshev, Jarrett Skirball, Krzysztof Kamocki, Edward Schuchman, Rubin M Tuder, Irina Petrache
ABSTRACT
The de novo pathway of ceramide synthesis has been implicated in the pathogenesis of excessive lung apoptosis and murine emphysema. Intracellular and paracellular-generated ceramides may trigger apoptosis and propagate the death signals to neighboring cells, respectively. In this study we compared the sphingolipid signaling pathways triggered by the paracellular- versus intracellular-generated ceramides as they induce lung endothelial cell apoptosis, a process important in emphysema development. Intermediate-chain length (C(8:0)) extracellular ceramides, used as a surrogate of paracellular ceramides, triggered caspase-3 activation in primary mouse lung endothelial cells, similar to TNF-alpha-generated endogenous ceramides. Inhibitory siRNA against serine palmitoyl transferase subunit 1 but not acid sphingomyelinase inhibited both C(8:0) ceramide- and TNF-alpha (plus cycloheximide)-induced apoptosis, consistent with the requirement for activation of the de novo pathway of sphingolipid synthesis. Tandem mass spectrometry analysis detected increases in both relative and absolute levels of C(16:0) ceramide in response to C(8:0) and TNF-alpha treatments. These results implicate the de novo pathway of ceramide synthesis in the apoptotic effects of both paracellular ceramides and TNF-alpha-stimulated intracellular ceramides in primary lung endothelial cells. The serine palmitoyl synthase-regulated ceramides synthesis may contribute to the amplification of pulmonary vascular injury induced by excessive ceramides.
Product Description
Avanti
C12-NBD Ceramide, Avanti Research - A Croda Brand 810211P, powder