MilliporeSigma
  • CWR22: the first human prostate cancer xenograft with strongly androgen-dependent and relapsed strains both in vivo and in soft agar.
CWR22: the first human prostate cancer xenograft with strongly androgen-dependent and relapsed strains both in vivo and in soft agar.
Cancer research (1996-07-01)
M Nagabhushan, C M Miller, T P Pretlow, J M Giaconia, N L Edgehouse, S Schwartz, H J Kung, R W de Vere White, P H Gumerlock, M I Resnick, S B Amini, T G Pretlow
ABSTRACT
Most patients' prostate cancers respond to androgen deprivation but relapse after periods of several months to years. Only two prostate cancer xenografts, LNCaP and PC-346, have been reported to be responsive to androgen deprivation and to relapse subsequently. Both of these tumors shrink slightly, if at all, and relapse less than 5 weeks after androgen withdrawal. After androgen withdrawal, the human primary prostate cancer xenograft CWR22 regresses markedly, and prostate-specific antigen (PSA) falls up to 3000-fold in the blood of mice. PSA usually returns to normal. In some animals, the tumor relapses and is then designated CWR22R. In these animals, PSA starts to rise approximately 2-7 months, and tumor begins to grow 3-10 months after castration. Animals with CWR22 need to be euthanized because of large tumors 6-12 weeks after the transplantation of CWR22. Androgen withdrawal prolongs life approximately 3-4-fold.
Sigma-Aldrich
CWR-R1ca Human Prostate Cancer Cell Line, CWR-R1ca Human Prostate Cancer Cell Line is a valuable cell model for castration-recurrent prostate cancer studies.