MilliporeSigma
Phosphatidic acid in membrane rearrangements.
FEBS letters (2019-08-01)
Mikhail A Zhukovsky, Angela Filograna, Alberto Luini, Daniela Corda, Carmen Valente
ABSTRACT
Phosphatidic acid (PA) is the simplest cellular glycerophospholipid characterized by unique biophysical properties: a small headgroup; negative charge; and a phosphomonoester group. Upon interaction with lysine or arginine, PA charge increases from -1 to -2 and this change stabilizes protein-lipid interactions. The biochemical properties of PA also allow interactions with lipids in several subcellular compartments. Based on this feature, PA is involved in the regulation and amplification of many cellular signalling pathways and functions, as well as in membrane rearrangements. Thereby, PA can influence membrane fusion and fission through four main mechanisms: it is a substrate for enzymes producing lipids (lysophosphatidic acid and diacylglycerol) that are involved in fission or fusion; it contributes to membrane rearrangements by generating negative membrane curvature; it interacts with proteins required for membrane fusion and fission; and it activates enzymes whose products are involved in membrane rearrangements. Here, we discuss the biophysical properties of PA in the context of the above four roles of PA in membrane fusion and fission.
Product Description
Avanti
16:0-06:0 NBD PA, Avanti Research - A Croda Brand 810173P, powder
Avanti
18:1-12:0 NBD PA, Avanti Research - A Croda Brand
Avanti
14:0-12:0 NBD PA, Avanti Research - A Croda Brand 810172P, powder
Avanti
16:0-12:0 NBD PA, Avanti Research - A Croda Brand 810174P, powder