MilliporeSigma
  • Synthetic sulfogalactosylceramide (sulfatide) and its use for the mass spectrometric quantitative urinary determination in metachromatic leukodystrophies.
Synthetic sulfogalactosylceramide (sulfatide) and its use for the mass spectrometric quantitative urinary determination in metachromatic leukodystrophies.
Glycoconjugate journal (2007-08-19)
Yanli Cui, Benoit Colsch, Carlos Afonso, Carlos Alonso, Nicole Baumann, Jean-Claude Tabet, Jean-Maurice Mallet, Yongmin Zhang
ABSTRACT
3-O-Sulfogalactosylceramides (sulfatides) accumulate in the genetic disease metachromatic leukodystrophy which is due to a defect in the catabolic enzyme, arylsulfatase A. Clinical diagnosis is usually confirmed by in vitro enzymatic deficiency of arylsulfatase A activity. The diagnosis may be complicated because of arylsulfatase A pseudo-deficiencies and another cause of MLD, sphingolipid activator B deficiency. As large quantities of sulfatides can be found in the urine in this disease, sulfatiduria appears as an extremely useful test. As recently enzyme replacement is underway, the quantitative determination, using an internal standard, appears particularly useful as a follow-up. Thus a non-physiological sulfatide was synthesized for this purpose, i.e. 3-O-sulfo-beta-D-C17 galactosylceramide (3-O-Sulfo-D: -Galactosyl-beta1'-->1-N-Heptadecanoyl-D-erythro-Sphingosine). It has been prepared through condensation of an azidosphingosine derivative with a protected D-galactopyranosyltrichloroacetimidate. Reduction of the azide was followed by acylation of a C-17 fatty acid. The key step was achieved by selective sulfation of the desired hydroxyl group on the sugar residue of the galactosylceramide using the stannylene methodology to give a 3'-sulfated beta-galactosyl C-17 ceramide.
Product Description
Avanti
C24:1 Mono-sulfo galactosyl (alpha) ceramide (d18:1/24:1), Avanti Research - A Croda Brand 860871P, powder