MilliporeSigma
  • Distinct APCs explain the cytokine bias of α-galactosylceramide variants in vivo.
Distinct APCs explain the cytokine bias of α-galactosylceramide variants in vivo.
Journal of immunology (Baltimore, Md. : 1950) (2012-03-07)
Li Bai, Michael G Constantinides, Seddon Y Thomas, Rachel Reboulet, Fanyong Meng, Frank Koentgen, Luc Teyton, Paul B Savage, Albert Bendelac
ABSTRACT
α-Galactosylceramide represents a new class of vaccine adjuvants and immunomodulators that stimulate NKT cells to secrete Th1 and Th2 cytokines. Synthetic variants with short or unsaturated acyl chains exhibit a striking Th2 bias in vivo but no evidence of defect in TCR signaling or stimulation of NKT cells in vitro. Using cd1d1(fl/fl) mice, we demonstrated that distinct APC types explained the cytokine bias in vivo. Whereas NKT stimulation by α-Galactosylceramide required CD1d expression by dendritic cells (DCs), presentation of the Th2 variants was promiscuous and unaffected by DC-specific ablation of CD1d. This DC-independent stimulation failed to activate the feedback loop between DC IL-12 and NK cell IFN-γ, explaining the Th2 bias. Conversely, forced presentation of the Th2 variants by DC induced high IL-12. Thus, lipid structural variations that do not alter TCR recognition can activate distinct Th1 or Th2 cellular networks by changing APC targeting in vivo.
Product Description
Avanti
C6-NBD Galactosyl Ceramide, Avanti Research - A Croda Brand 810220P, powder