MilliporeSigma
  • Puromycin-sensitive alanyl aminopeptidase from human liver cytosol: purification and characterization.
Puromycin-sensitive alanyl aminopeptidase from human liver cytosol: purification and characterization.
Forensic science international (2000-09-09)
Y Yamamoto, Y H Li, I Ushiyama, A Nishimura, I Ohkubo, K Nishi
ABSTRACT
A cytosolic alanyl aminopeptidase (AAP-S) was purified to homogeneity from human liver cytosol. The molecular weight of the purified enzyme was calculated to be approximately 98,000 on TOF-MS and 90,000 on SDS-PAGE in the presence of beta-ME. These findings suggest that the enzyme exists as a monomeric form in human liver cytosol. The enzyme rapidly hydrolyzed the substrates Ala-, Lys- and Phe-MCAs, and moderately hydrolyzed Met-, Leu-, Tyr- and Lys-Ala-MCAs at pH ranging from 7.5 to 8.0. The order of the K(cat)/K(m) values of AAP-S at the optimal pH was Arg->Arg-Arg->Met->Leu->Lys->Phe->Lys-Ala->Tyr->Ala-MCAs. It was strongly inhibited by bestatin, leuhistin, actinonin, amastatin, 1, 10-phenanthroline, DFP, PCMBS, Zn(2+), Cd(2+), Co(2+), Cu((2+)), Hg(2+) and puromycin. AAP-S was approximately 80 times more sensitive than human seminal plasma AAP (aminopeptidase N, membrane type). The amino acid sequence of the first 60 residues of AAP-S was highly homologous with the N-terminal amino acid sequence of the rat liver puromycin-sensitive enkephalin-degrading aminopeptidase. These physicochemical properties and findings indicate that AAP-S from human liver cytosol is identical to those of other puromycin-sensitive aminopeptidase(s). Furthermore, with immunohistochemistry the enzyme was strongly stained in the cytoplasm of liver cells and renal tubules, and was ubiquitously localized in various human tissues.
Sigma-Aldrich
Puromycin ready made solution, 10 mg/mL in H2O, ≥98% (HPLC), Cell culture tested