MilliporeSigma
  • Discovery of a structurally novel opioid kappa-agonist derived from 4,5-epoxymorphinan.
Discovery of a structurally novel opioid kappa-agonist derived from 4,5-epoxymorphinan.
Chemical & pharmaceutical bulletin (1998-03-21)
H Nagase, J Hayakawa, K Kawamura, K Kawai, Y Takezawa, H Matsuura, C Tajima, T Endo
ABSTRACT
A new type of kappa-agonist, 17-cyclopropylmethyl-3, 14 beta-dihydroxy-4,5 alpha-epoxy-6 beta-[N-methyl-trans-3-(3-furyl) acrylamido]morphinan hydrochloride (1, TRK-820), was discovered by a new working hypothesis. The "message-address concept" for opioid antagonists and the "accessory site" for general antagonists were applied to design TRK-820. A unique structural feature of TRK-820, which is different from other prototypical kappa-opioid receptor agonists, is the existence of the 4,5-epoxymorphinan structure with a tyrosine-glysine moiety for endogenous opioid peptides such as dynorphins. TRK-820 exhibited high potency and high kappa-selectivity in guinea pig ileum (GPI) and mouse vas deferens (MVD) preparations. In the mouse acetic-acid-induced writhing model and mouse tail flick model of antinociception, TRK-820 was 85-140 times more potent than morphine and 85-350 times more potent than U-50488H. This structurally novel kappa-agonist showed neither aversion nor preference in the Conditioned Place Preference test, in spite of the fact that prototypes of kappa-agonists (U-50488H derivatives) demonstrated aversion.